siRNA Details
Gene Symbol: LOC284581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046098.1 | 1 | 385 | |||
GenBank mRNA | AK094426.1 | - | 1 | 870 | ||
AK096315.1 | - | 1 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
