siRNA Details
Gene Symbol: SFMBT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018039.1 | NP_001018049.1 | 4, 5 | 534 | ||
NM_001029880.2 | NP_001025051.1 | 4, 5 | 636 | |||
XM_005252551.2 | XP_005252608.1 | 4, 5 | 608 | |||
XM_011519611.1 | XP_011517913.1 | 4, 5 | 635 | |||
XM_017016466.1 | XP_016871955.1 | 4, 5 | 537 | |||
GenBank mRNA | AB046837.1 | - | 4, 5 | 534 | ||
AK090887.1 | - | 4, 5 | 642 | |||
BC144327.1 | - | 4, 5 | 601 | |||
BC152430.1 | - | 4, 5 | 534 | 8862546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
