siRNA Details
Gene Symbol: PEG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040152.1 | NP_001035242.1 | 2 | 6194 | ||
NM_001172437.2 | NP_001165908.1 | 2 | 6205 | |||
NM_001172438.2 | NP_001165909.1 | 2 | 6205 | |||
NM_001184961.1 | NP_001171890.1 | 2 | 6194 | |||
NM_001184962.1 | NP_001171891.1 | 2 | 6194 | |||
NM_015068.3 | NP_055883.2 | 2 | 6194 | |||
GenBank mRNA | AB028974.1 | - | 1 | 5767 | ||
AB049150.1 | - | 2 | 5978 | |||
AB049834.1 | - | 1 | 5832 | |||
AF038197.1 | - | 1 | 1127 | |||
AF216076.1 | - | 1 | 5724 | |||
AK025529.1 | - | 1 | 3529 | |||
BC015448.1 | - | 1 | 1608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
