siRNA Details
Gene Symbol: C1orf68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024679.2 | NP_001019850.1 | 1 | 761 | ||
GenBank mRNA | CU446285.1 | - | 1 | 601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
