siRNA Details
Gene Symbol: SNRPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004597.6 | NP_004588.1 | 3 | 337 | ||
NM_177542.3 | NP_808210.2 | 3 | 301 | |||
GenBank mRNA | AJ707552.1 | - | 1 | 22 | ||
AK291912.1 | - | 2 | 116 | |||
BC000486.2 | - | 2 | 108 | 2819767 | ||
BC001930.2 | - | 1 | 86 | 2819767 | ||
BG528418.1 | - | 232 | ||||
BU531743.1 | - | 3 | 226 | |||
BU532756.1 | - | 382 | ||||
CB136390.1 | - | 2 | 527 | |||
CF130080.1 | - | 3 | 337 | |||
DA325073.1 | - | 3 | 229 | |||
U15008.1 | - | 2 | 112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
