siRNA Details
Gene Symbol: SNRPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004597.6 | NP_004588.1 | 4 | 531 | ||
NM_177542.3 | NP_808210.2 | 4 | 495 | |||
GenBank mRNA | AJ707552.1 | - | 2 | 216 | ||
AK291912.1 | - | 3 | 310 | |||
BC000486.2 | - | 3 | 302 | 2819767 | ||
BC001930.2 | - | 2 | 280 | 2819767 | ||
BG528418.1 | - | 426 | ||||
BU531743.1 | - | 4 | 420 | |||
DA325073.1 | - | 4 | 423 | |||
U15008.1 | - | 3 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
