siRNA Details
Gene Symbol: LGALS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002308.3 | NP_002299.2 | 2 | 185 | ||
NM_009587.2 | NP_033665.1 | 2 | 185 | |||
NR_024043.1 | 2 | 185 | ||||
XM_006721892.2 | XP_006721955.1 | 2 | 148 | |||
XM_006721893.2 | XP_006721956.1 | 2 | 177 | |||
XM_006721894.3 | XP_006721957.1 | 2 | 148 | |||
XM_006721895.3 | XP_006721958.1 | 2 | 148 | |||
XM_011524796.1 | XP_011523098.1 | 2 | 177 | |||
XM_017024623.1 | XP_016880112.1 | 2 | 177 | |||
GenBank mRNA | AB003517.1 | - | 2 | 92 | ||
AB005894.1 | - | 2 | 126 | |||
AB006782.1 | - | 2 | 148 | |||
AB008492.1 | - | 2 | 92 | |||
AK126017.1 | - | 2 | 1535 | |||
AK223232.1 | - | 2 | 149 | |||
AK296031.1 | - | 2 | 127 | |||
AK301624.1 | - | 2 | 126 | |||
BC105942.1 | - | 2 | 109 | 40031795 | ||
BC105944.1 | - | 2 | 109 | 40031802 | ||
BC110340.1 | - | 2 | 126 | 5760999 | ||
Z49107.1 | - | 2 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
