siRNA Details
Gene Symbol: LGALS9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040078.2 | NP_001035167.2 | 11 | 1693 | ||
XM_005256769.1 | XP_005256826.1 | 11 | 1690 | |||
XM_005256770.1 | XP_005256827.1 | 10 | 1594 | |||
XM_011523993.1 | XP_011522295.1 | 10 | 1597 | |||
XM_011523994.1 | XP_011522296.1 | 9 | 1561 | |||
XM_011523995.1 | XP_011522297.1 | 11 | 1605 | |||
XM_011523996.1 | XP_011522298.1 | 10 | 1597 | |||
XM_011523997.1 | XP_011522299.1 | 9 | 1331 | |||
GenBank mRNA | BC073889.1 | - | 11 | 1662 | 5222492 |
Gene Symbol: LGALS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002308.3 | NP_002299.2 | 10 | 1642 | ||
NM_009587.2 | NP_033665.1 | 11 | 1738 | |||
NR_024043.1 | 10 | 1571 | ||||
XM_006721892.2 | XP_006721955.1 | 9 | 1569 | |||
XM_006721893.2 | XP_006721956.1 | 10 | 1567 | |||
XM_006721894.3 | XP_006721957.1 | 9 | 1442 | |||
XM_006721895.3 | XP_006721958.1 | 8 | 1406 | |||
XM_017024623.1 | XP_016880112.1 | 10 | 1694 | |||
GenBank mRNA | AB005894.1 | - | 10 | 1583 | ||
AB006782.1 | - | 11 | 1701 | |||
AK126017.1 | - | 10 | 2992 | |||
AK223232.1 | - | 10 | 1606 | |||
AK301624.1 | - | 10 | 1445 | |||
BC110340.1 | - | 10 | 1583 | 5760999 | ||
Z49107.1 | - | 10 | 1611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
