siRNA Details
Gene Symbol: KIR3DX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026716.2 | 3, 4 | 463 | |||
NR_136268.1 | 3, 4 | 463 | ||||
GenBank mRNA | AK024467.1 | - | 1 | 316 | ||
BC171834.1 | - | 2, 3 | 673 | |||
DQ224421.1 | - | 3, 4 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
