siRNA Details
Gene Symbol: SLC5A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011547.2 | NP_001011547.2 | 9 | 1170 | ||
NM_001135181.1 | NP_001128653.1 | 10 | 1245 | |||
XM_011540924.2 | XP_011539226.1 | 9 | 2851 | |||
XM_011540925.2 | XP_011539227.1 | 9 | 2930 | |||
XM_011540926.2 | XP_011539228.1 | 8 | 1431 | |||
XM_011540927.1 | XP_011539229.1 | 7 | 1029 | |||
XM_011540928.1 | XP_011539230.1 | 9 | 1049 | |||
XM_011540929.2 | XP_011539231.1 | 9 | 2934 | |||
XM_017000558.1 | XP_016856047.1 | 9 | 2933 | |||
GenBank mRNA | AK131200.1 | - | 9 | 1123 | ||
AK296133.1 | - | 8 | 843 | |||
BC104857.1 | - | 9 | 1170 | 8143860 | ||
BC104863.1 | - | 9 | 1170 | 8143866 | ||
BC143570.1 | - | 10 | 1233 | 9052081 | ||
BC143572.1 | - | 10 | 1243 | 9052083 | ||
BC143587.1 | - | 8 | 964 | |||
BX648549.1 | - | 10 | 1199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
