siRNA Details
Gene Symbol: SLC5A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011547.2 | NP_001011547.2 | 7 | 904 | ||
NM_001135181.1 | NP_001128653.1 | 8 | 979 | |||
XM_011540925.2 | XP_011539227.1 | 7 | 2664 | |||
XM_011540926.2 | XP_011539228.1 | 6 | 1165 | |||
XM_011540927.1 | XP_011539229.1 | 5 | 763 | |||
XM_011540928.1 | XP_011539230.1 | 7 | 783 | |||
XM_011540929.2 | XP_011539231.1 | 7 | 2668 | |||
XM_017000558.1 | XP_016856047.1 | 7 | 2667 | |||
GenBank mRNA | AK131200.1 | - | 7 | 857 | ||
BC104857.1 | - | 7 | 904 | 8143860 | ||
BC104863.1 | - | 7 | 904 | 8143866 | ||
BC143570.1 | - | 8 | 967 | 9052081 | ||
BC143572.1 | - | 8 | 977 | 9052083 | ||
BX648549.1 | - | 8 | 933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
