siRNA Details
Gene Symbol: FAM149B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173348.1 | NP_775483.1 | 14 | 2196 | ||
XM_005269744.2 | XP_005269801.1 | 14 | 2172 | |||
XM_005269745.2 | XP_005269802.1 | 13 | 2070 | |||
XM_005269747.3 | XP_005269804.1 | 14 | 1987 | |||
XM_011539737.2 | XP_011538039.1 | 14 | 1999 | |||
XM_017016164.1 | XP_016871653.1 | 13 | 1874 | |||
XM_017016165.1 | XP_016871654.1 | 13 | 1843 | |||
XM_017016166.1 | XP_016871655.1 | 13 | 1880 | |||
GenBank mRNA | AB023191.1 | - | 13 | 1924 | ||
AK023439.1 | - | 10 | 1255 | |||
AK025189.1 | - | 3 | 1584 | |||
AK303436.1 | - | 14 | 1960 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
