siRNA Details
Gene Symbol: PHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004426.2 | NP_004417.2 | 2 | 4713 | ||
XM_005253334.2 | XP_005253391.1 | 15 | 4713 | |||
XM_011520599.2 | XP_011518901.1 | 15 | 4716 | |||
XM_011520600.2 | XP_011518902.1 | 14 | 4590 | |||
XM_011520603.2 | XP_011518905.1 | 15 | 4820 | |||
XM_017018955.1 | XP_016874444.1 | 15 | 5108 | |||
XM_017018956.1 | XP_016874445.1 | 14 | 4566 | |||
XM_017018957.1 | XP_016874446.1 | 14 | 4539 | |||
XM_017018958.1 | XP_016874447.1 | 14 | 4515 | |||
GenBank mRNA | BC073964.1 | - | 3 | 4562 | 5788132 | |
BX640639.1 | - | 18 | 3403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
