siRNA Details
Gene Symbol: NDST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268441.1 | XP_005268498.1 | 7 | 2370 | ||
GenBank mRNA | BC012888.1 | - | 6 | 2115 | 3882074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
