siRNA Details
Gene Symbol: FAM214A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286495.1 | NP_001273424.1 | 5 | 1635 | ||
NM_019600.3 | NP_062546.2 | 6 | 1634 | |||
NR_104457.1 | 6 | 1776 | ||||
XM_005254547.2 | XP_005254604.1 | 6 | 1597 | |||
XM_005254548.2 | XP_005254605.1 | 6 | 2116 | |||
XM_011521789.2 | XP_011520091.1 | 6 | 1663 | |||
XM_011521790.1 | XP_011520092.1 | 5 | 1635 | |||
GenBank mRNA | AK292917.1 | - | 6 | 1623 | ||
AK293920.1 | - | 5 | 1625 | |||
AK308049.1 | - | 5 | 1440 | |||
BC109128.2 | - | 5 | 1565 | 40036502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
