siRNA Details
Gene Symbol: TTC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310135.1 | NP_001297064.1 | 33 | 6624 | ||
XM_011537430.2 | XP_011535732.1 | 29 | 4456 | |||
XM_011537431.2 | XP_011535733.1 | 27 | 4113 | |||
XM_017021254.1 | XP_016876743.1 | 32 | 6474 | |||
XM_017021255.1 | XP_016876744.1 | 31 | 6339 | |||
XM_017021256.1 | XP_016876745.1 | 32 | 6738 | |||
GenBank mRNA | BC014342.2 | - | 6 | 1031 | ||
BX161415.1 | - | 13 | 1908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
