siRNA Details
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136214.2 | NP_001129686.1 | 13 | 2082 | ||
NM_001271197.1 | NP_001258126.1 | 13 | 2162 | |||
NM_182619.3 | NP_872425.2 | 13 | 2188 | |||
GenBank mRNA | BC008616.1 | - | 3 | 1076 |
Gene Symbol: CLEC18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006721179.1 | XP_006721242.1 | 12 | 2146 | ||
XM_011523034.1 | XP_011521336.1 | 13 | 2392 | |||
XM_011523036.2 | XP_011521338.1 | 11 | 1976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
