siRNA Details
Gene Symbol: WTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080436.1 | NP_001073905.1 | 8 | 1455 | ||
GenBank mRNA | AK130059.1 | - | 7 | 793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
