siRNA Details
Gene Symbol: FAM160B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020940.3 | NP_065991.3 | 17 | 4202 | ||
GenBank mRNA | AB046820.2 | - | 16 | 3797 | ||
AL833226.1 | - | 6 | 2261 | |||
CR933692.1 | - | 17 | 3922 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
