siRNA Details
Gene Symbol: VPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009921.2 | NP_001009921.1 | 16 | 1408 | ||
NM_015303.3 | NP_056118.2 | 15 | 1402 | |||
XM_005247251.3 | XP_005247308.1 | 16 | 1676 | |||
XM_005247253.4 | XP_005247310.1 | 16 | 1414 | |||
XM_006713556.3 | XP_006713619.1 | 8 | 696 | |||
XM_011512599.2 | XP_011510901.1 | 3 | 158 | |||
XM_011512600.2 | XP_011510902.1 | 8 | 610 | |||
XM_011512601.2 | XP_011510903.1 | 8 | 696 | |||
XM_017006040.1 | XP_016861529.1 | 14 | 1096 | |||
XM_017006041.1 | XP_016861530.1 | 16 | 1414 | |||
GenBank mRNA | AB018347.1 | - | 8 | 587 | ||
AK056661.1 | - | 16 | 1380 | |||
AK292423.1 | - | 15 | 1374 | |||
AL833838.1 | - | 13 | 941 | |||
BC140768.1 | - | 16 | 1344 | 9021636 | ||
BX647915.1 | - | 15 | 1363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
