siRNA Details
Gene Symbol: VPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009921.2 | NP_001009921.1 | 19 | 1710 | ||
NM_015303.3 | NP_056118.2 | 18 | 1704 | |||
XM_005247251.3 | XP_005247308.1 | 19 | 1978 | |||
XM_005247253.4 | XP_005247310.1 | 19 | 1716 | |||
XM_006713556.3 | XP_006713619.1 | 11 | 998 | |||
XM_011512599.2 | XP_011510901.1 | 6 | 460 | |||
XM_011512600.2 | XP_011510902.1 | 11 | 912 | |||
XM_011512601.2 | XP_011510903.1 | 11 | 998 | |||
XM_017006040.1 | XP_016861529.1 | 17 | 1398 | |||
XM_017006041.1 | XP_016861530.1 | 19 | 1716 | |||
GenBank mRNA | AB018347.1 | - | 11 | 889 | ||
AK056661.1 | - | 19 | 1682 | |||
AK292423.1 | - | 18 | 1676 | |||
AL833838.1 | - | 16 | 1243 | |||
BC140768.1 | - | 19 | 1646 | 9021636 | ||
BX647915.1 | - | 18 | 1665 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
