siRNA Details
Gene Symbol: ANKRD36BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015424.1 | 5, 6 | 535 | |||
GenBank mRNA | AL137717.1 | - | 4, 5 | 473 | ||
BC031698.1 | - | 5, 6 | 535 | |||
BX648045.1 | - | 5, 6 | 569 |
Gene Symbol: ANKRD36C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310154.1 | NP_001297083.1 | 26, 27 | 2442 | ||
GenBank mRNA | AK302433.1 | - | 25, 26 | 1843 | ||
AL832836.1 | - | 7, 8 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
