siRNA Details
Gene Symbol: UVSSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317934.1 | NP_001304863.1 | 12 | 2118 | ||
NM_001317935.1 | NP_001304864.1 | 12 | 2106 | |||
NM_020894.3 | NP_065945.2 | 12 | 2219 | |||
XM_017008490.1 | XP_016863979.1 | 12 | 1940 | |||
XM_017008491.1 | XP_016863980.1 | 12 | 2032 | |||
XM_017008492.1 | XP_016863981.1 | 12 | 2030 | |||
XM_017008493.1 | XP_016863982.1 | 12 | 1906 | |||
XM_017008494.1 | XP_016863983.1 | 12 | 1791 | |||
XM_017008495.1 | XP_016863984.1 | 9 | 1933 | |||
XM_017008500.1 | XP_016863989.1 | 6 | 660 | |||
GenBank mRNA | AL390128.1 | - | 4 | 446 | ||
BC021930.1 | - | 5 | 857 | |||
EL735271.1 | - | 4 | 425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
