siRNA Details
Gene Symbol: GPX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008397.3 | NP_001008398.2 | 3 | 1696 | ||
NM_001306197.1 | NP_001293126.1 | 3 | 1543 | |||
NM_001306198.1 | NP_001293127.1 | 2 | 1434 | |||
NM_001306201.1 | NP_001293130.1 | 3 | 1417 | |||
NR_131336.1 | 4 | 1610 | ||||
NR_131337.1 | 4 | 1552 | ||||
NR_131338.1 | 3 | 1348 | ||||
NR_131340.1 | 3 | 1686 | ||||
XM_006714631.2 | XP_006714694.1 | 3 | 1662 | |||
GenBank mRNA | BX537894.1 | - | 4 | 1560 | ||
BX647770.1 | - | 2 | 1384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
