siRNA Details
Gene Symbol: OTUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145373.2 | NP_001138845.1 | 1 | 1480 | ||
GenBank mRNA | AB188491.1 | - | 1 | 1480 | ||
AK096389.1 | - | 1 | 325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
