siRNA Details
Gene Symbol: C16orf92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109659.1 | NP_001103129.1 | 4 | 557 | ||
NM_001109660.1 | NP_001103130.1 | 4 | 637 | |||
GenBank mRNA | AK058133.1 | - | 4 | 637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
