siRNA Details
Gene Symbol: C8orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256596.1 | NP_001243525.1 | 5 | 372 | ||
NM_001256597.1 | NP_001243526.1 | 5 | 480 | |||
NM_001256598.1 | NP_001243527.1 | 5 | 1122 | |||
GenBank mRNA | AJ276240.1 | - | 5 | 1109 | ||
BG202040.1 | - | 5 | 398 | |||
DA900243.1 | - | 5 | 480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
