siRNA Details
Gene Symbol: LMOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207163.1 | NP_997046.1 | 1 | 364 | ||
GenBank mRNA | AK299389.1 | - | 1 | 305 | ||
AK299434.1 | - | 1 | 308 | |||
AK300698.1 | - | 1 | 305 | |||
AK309496.1 | - | 1 | 302 | |||
BC017911.1 | - | 1 | 305 | |||
BC062765.1 | - | 1 | 310 | |||
DC385641.1 | - | 1 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
