siRNA Details
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128933.2 | NP_001122405.1 | 4 | 4997 | ||
GenBank mRNA | AJ010482.1 | - | 2 | 4023 | ||
AL832031.1 | - | 4 | 4986 | |||
AL832363.1 | - | 4 | 4897 | |||
AL833294.1 | - | 4 | 4983 | |||
BX647849.1 | - | 2 | 4512 | |||
FN422000.1 | - | 2 | 4058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
