siRNA Details
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128933.2 | NP_001122405.1 | 4 | 6474 | ||
GenBank mRNA | AL833294.1 | - | 4 | 6459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
