siRNA Details
Gene Symbol: DANT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130730.1 | 1 | 109 | |||
NR_132337.1 | 1 | 109 | ||||
GenBank mRNA | BC003645.1 | - | 1 | 87 | ||
BE297956.1 | - | 1 | 110 | |||
BM925596.1 | - | 1 | 131 | |||
BX642309.1 | - | 1 | 116 | |||
KM192214.1 | - | 11 | ||||
KM192215.1 | - | 11 | ||||
KM192216.1 | - | 11 | ||||
KM192217.1 | - | 11 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
