siRNA Details
Gene Symbol: ZBTB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040441.2 | NP_001035531.2 | 5 | 1830 | ||
NM_001291496.1 | NP_001278425.1 | 5 | 1649 | |||
NR_111980.1 | 4 | 825 | ||||
GenBank mRNA | AF548353.1 | - | 4 | 1362 | ||
AK074546.1 | - | 5 | 1548 | |||
AY157873.1 | - | 5 | 1365 | |||
BC015067.1 | - | 3 | 704 | |||
BC015239.1 | - | 5 | 1498 | 3914043 | ||
BC029654.1 | - | 3 | 750 | |||
BQ723271.1 | - | 3 | 342 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
