siRNA Details
Gene Symbol: KIAA0226L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286763.1 | NP_001273692.1 | 2 | 419 | ||
NM_001286765.1 | NP_001273694.1 | 2 | 419 | |||
NM_025113.3 | NP_079389.2 | 2 | 419 | |||
GenBank mRNA | AK125950.1 | - | 3 | 1061 | ||
AK300183.1 | - | 2 | 123 | |||
AK310264.1 | - | 2 | 313 | |||
BC043488.1 | - | 2 | 152 | 5205680 | ||
BX401854.2 | - | 2 | 153 | |||
DB151884.1 | - | 2 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
