siRNA Details
Gene Symbol: TMEM64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008495.3 | NP_001008495.2 | 3 | 2581 | ||
NM_001146273.1 | NP_001139745.1 | 2 | 2425 | |||
GenBank mRNA | AK095472.1 | - | 1 | 792 | ||
AK123958.1 | - | 3 | 2276 | |||
AL834364.1 | - | 3 | 2286 | |||
BC017409.1 | - | 1 | 478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
