siRNA Details
Gene Symbol: IQSEC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111125.2 | NP_001104595.1 | 14, 15 | 3703 | ||
XM_006724579.2 | XP_006724642.1 | 15, 16 | 3825 | |||
XM_006724580.3 | XP_006724643.1 | 14, 15 | 2993 | |||
XM_011530773.2 | XP_011529075.1 | 13, 14 | 2897 | |||
XM_017029359.1 | XP_016884848.1 | 13, 14 | 3699 | |||
XM_017029360.1 | XP_016884849.1 | 14, 15 | 3111 | |||
GenBank mRNA | AB011094.1 | - | 14, 15 | 3703 | ||
FJ154892.1 | - | 2, 3 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
