siRNA Details
Gene Symbol: FAXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032385.4 | NP_115761.2 | 6, 7 | 772 | ||
XM_005268358.2 | XP_005268415.1 | 3, 4 | 417 | |||
XM_006714753.2 | XP_006714816.1 | 7, 8 | 1147 | |||
XM_017008965.1 | XP_016864454.1 | 7, 8 | 1172 | |||
XM_017008966.1 | XP_016864455.1 | 4, 5 | 447 | |||
GenBank mRNA | AK127944.1 | - | 1, 2 | 1319 | ||
AK295546.1 | - | 5, 6 | 649 | |||
BC004506.2 | - | 4, 5 | 469 | |||
BC007216.1 | - | 6, 7 | 740 | 2906244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
