siRNA Details
Gene Symbol: MECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016011.3 | NP_057095.3 | 1, 2 | 228 | ||
XM_011541540.1 | XP_011539842.1 | 1, 2 | 217 | |||
GenBank mRNA | AF151821.1 | - | 1, 2 | 202 | ||
AK025830.1 | - | 1, 2 | 194 | |||
AK225687.1 | - | 1, 2 | 194 | |||
AK300889.1 | - | 1, 2 | 192 | |||
AK315274.1 | - | 1, 2 | 215 | |||
BC001419.2 | - | 1, 2 | 199 | 3139846 | ||
CR456703.1 | - | 1, 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
