siRNA Details
Gene Symbol: HMGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319077.1 | NP_001306006.1 | 6 | 1828 | ||
NM_001319078.1 | NP_001306007.1 | 5 | 1749 | |||
NM_001319079.1 | NP_001306008.1 | 6 | 1861 | |||
NM_001319080.1 | NP_001306009.1 | 6 | 1860 | |||
NM_001319081.1 | NP_001306010.1 | 5 | 1747 | |||
NM_001319082.1 | NP_001306011.1 | 6 | 1883 | |||
NM_002131.3 | NP_002122.1 | 6 | 1830 | |||
NM_145899.2 | NP_665906.1 | 6 | 1863 | |||
NM_145901.2 | NP_665908.1 | 5 | 2030 | |||
NM_145902.2 | NP_665909.1 | 5 | 1997 | |||
NM_145903.2 | NP_665910.1 | 5 | 1716 | |||
NM_145905.2 | NP_665912.1 | 5 | 1719 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
