siRNA Details
Gene Symbol: PHF20L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016018.4 | NP_057102.4 | 6, 7 | 840 | ||
XM_005250931.1 | XP_005250988.1 | 6, 7 | 822 | |||
XM_011517075.1 | XP_011515377.1 | 6, 7 | 822 | |||
XM_011517076.1 | XP_011515378.1 | 6, 7 | 818 | |||
XM_011517077.1 | XP_011515379.1 | 6, 7 | 822 | |||
XM_011517078.1 | XP_011515380.1 | 6, 7 | 810 | |||
XM_011517079.1 | XP_011515381.1 | 6, 7 | 810 | |||
XM_011517088.1 | XP_011515390.1 | 6, 7 | 822 | |||
XM_017013509.1 | XP_016868998.1 | 6, 7 | 806 | |||
XM_017013513.1 | XP_016869002.1 | 6, 7 | 3645 | |||
XM_017013514.1 | XP_016869003.1 | 6, 7 | 3641 | |||
XM_017013516.1 | XP_016869005.1 | 6, 7 | 810 | |||
GenBank mRNA | AB101203.1 | - | 6, 7 | 807 | ||
AF151830.1 | - | 3, 4 | 194 | |||
AK023711.1 | - | 1 | 285 | |||
BC002822.2 | - | 6, 7 | 798 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
