siRNA Details
Gene Symbol: YIPF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182592.2 | NP_872398.2 | 6 | 831 | ||
XM_011513679.2 | XP_011511981.1 | 7 | 6172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
