siRNA Details
Gene Symbol: S100A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024210.1 | NP_001019381.1 | 5 | 862 | ||
NM_001024211.1 | NP_001019382.1 | 3 | 578 | |||
NM_001024212.1 | NP_001019383.1 | 3 | 549 | |||
NM_001024213.1 | NP_001019384.1 | 3 | 517 | |||
NM_005979.2 | NP_005970.1 | 4 | 654 | |||
XM_005245434.3 | XP_005245491.1 | 5 | 769 | |||
XM_011509862.2 | XP_011508164.1 | 7 | 1069 | |||
XM_011509863.2 | XP_011508165.1 | 7 | 1036 | |||
XM_011509864.1 | XP_011508166.1 | 5 | 859 | |||
XM_017002034.1 | XP_016857523.1 | 6 | 952 | |||
XM_017002035.1 | XP_016857524.1 | 6 | 953 | |||
XM_017002036.1 | XP_016857525.1 | 3 | 686 | |||
GenBank mRNA | AK097132.1 | - | 5 | 2158 | ||
BC000632.2 | - | 2 | 427 | 3343203 | ||
BC068064.1 | - | 2 | 413 | 6074799 | ||
BC070291.1 | - | 4 | 654 | 6713909 | ||
BM762088.1 | - | 3 | 551 | |||
X99920.1 | - | 3 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
