siRNA Details
Gene Symbol: LOC494141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026563.1 | 2 | 1415 | |||
NR_026564.1 | 3 | 1270 | ||||
GenBank mRNA | AK131059.1 | - | 3 | 1248 | ||
BC026079.1 | - | 2 | 1422 | |||
BC034967.1 | - | 2 | 1422 | |||
BC044592.1 | - | 2 | 1415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
