siRNA Details
Gene Symbol: NOP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256539.3 | NP_001243468.2 | 5 | 846 | ||
NM_001256540.3 | NP_001243469.2 | 5 | 843 | |||
NM_001291305.2 | NP_001278234.1 | 5 | 877 | |||
NM_001291307.2 | NP_001278236.1 | 5 | 814 | |||
NM_001291308.2 | NP_001278237.1 | 4 | 773 | |||
NM_001317975.1 | NP_001304904.1 | 4 | 1023 | |||
NM_016391.7 | NP_057475.2 | 5 | 934 | |||
NR_134316.1 | 4 | 1040 | ||||
NR_134318.1 | 6 | 933 | ||||
XM_011534567.1 | XP_011532869.1 | 4 | 543 | |||
GenBank mRNA | AF151019.1 | - | 5 | 929 | ||
AF161460.1 | - | 5 | 608 | |||
AK300883.1 | - | 5 | 522 | |||
BC019331.1 | - | 5 | 678 | 3534019 | ||
BC032424.2 | - | 5 | 489 | 5416724 | ||
BC040106.1 | - | 5 | 594 | 6066208 | ||
BM826387.1 | - | 4 | 432 | |||
CB122385.1 | - | 5 | 506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
