siRNA Details
Gene Symbol: C16orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080524.1 | NP_001073993.1 | 2 | 350 | ||
XM_005255502.3 | XP_005255559.1 | 2 | 416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
