siRNA Details
Gene Symbol: NECAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005260510.1 | XP_005260567.1 | 12 | 1199 | ||
XM_011528991.1 | XP_011527293.1 | 9 | 909 | |||
XM_017028015.1 | XP_016883504.1 | 9 | 908 | |||
XM_017028016.1 | XP_016883505.1 | 9 | 837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
