siRNA Details
Gene Symbol: TMEM205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145416.1 | NP_001138888.1 | 4 | 780 | ||
NM_001321112.1 | NP_001308041.1 | 4 | 889 | |||
NM_001321113.1 | NP_001308042.1 | 4 | 699 | |||
NM_001321114.1 | NP_001308043.1 | 4 | 808 | |||
NM_033408.3 | NP_212133.1 | 4 | 899 | |||
NM_198536.2 | NP_940938.1 | 3 | 1278 | |||
XM_005259899.2 | XP_005259956.1 | 4 | 985 | |||
XM_017026769.1 | XP_016882258.1 | 2 | 590 | |||
GenBank mRNA | AK127147.1 | - | 3 | 3165 | ||
AK291979.1 | - | 4 | 759 | |||
AY358949.1 | - | 4 | 720 | |||
BC064948.1 | - | 3 | 1019 | 6025162 | ||
BC091472.1 | - | 4 | 707 | 30335220 | ||
CB994930.1 | - | 4 | 742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
