siRNA Details
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 28 | 4720 | ||
XM_005261909.1 | XP_005261966.1 | 29 | 4768 | |||
XM_005261910.2 | XP_005261967.1 | 29 | 4779 | |||
XM_005261911.4 | XP_005261968.1 | 28 | 4718 | |||
XM_006724413.2 | XP_006724476.1 | 30 | 4838 | |||
XM_011530682.2 | XP_011528984.1 | 29 | 4919 | |||
XM_017028703.1 | XP_016884192.1 | 30 | 4819 | |||
XM_017028704.1 | XP_016884193.1 | 29 | 4772 | |||
GenBank mRNA | AB002313.1 | - | 27 | 4637 | ||
AB075511.1 | - | 13 | 1830 | |||
AK025415.1 | - | 3 | 307 | |||
AK025701.1 | - | 15 | 2088 | |||
AK056543.1 | - | 11 | 1889 | |||
AK074932.1 | - | 10 | 1356 | |||
AK123131.1 | - | 1 | 225 | |||
AK126394.1 | - | 9 | 1429 | |||
BC004542.2 | - | 5 | 539 | |||
BT006887.1 | - | 5 | 523 | |||
S76730.1 | - | 6 | 870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
