siRNA Details
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 31 | 5093 | ||
XM_005261909.1 | XP_005261966.1 | 32 | 5141 | |||
XM_005261910.2 | XP_005261967.1 | 32 | 5152 | |||
XM_005261911.4 | XP_005261968.1 | 31 | 5091 | |||
XM_006724413.2 | XP_006724476.1 | 33 | 5211 | |||
XM_011530682.2 | XP_011528984.1 | 32 | 5292 | |||
XM_017028703.1 | XP_016884192.1 | 33 | 5192 | |||
XM_017028704.1 | XP_016884193.1 | 32 | 5145 | |||
GenBank mRNA | AB002313.1 | - | 30 | 5010 | ||
AK025415.1 | - | 6 | 680 | |||
AK025701.1 | - | 18 | 2461 | |||
AK056543.1 | - | 14 | 2262 | |||
AK074932.1 | - | 13 | 1729 | |||
AK123131.1 | - | 3 | 674 | |||
AK126394.1 | - | 12 | 1802 | |||
BC004542.2 | - | 8 | 912 | |||
BC110366.1 | - | 3 | 330 | |||
BT006887.1 | - | 8 | 896 | |||
S76730.1 | - | 9 | 1242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
