siRNA Details
Gene Symbol: KRTAP2-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165252.1 | NP_001158724.1 | 1 | 519 | ||
GenBank mRNA | BC063625.1 | - | 1 | 519 | 3907481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
