siRNA Details
Gene Symbol: ERV3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007253.3 | NP_001007254.2 | 2 | 1349 | ||
GenBank mRNA | AK074464.1 | - | 1 | 491 | ||
AK295189.1 | - | 2 | 1256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
